Anti-GRIN2A Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
XPS-2024
PRSIXPS-2024EA 686 EUR InStock
PRSIXPS-2024
Anti-GRIN2A Rabbit Polyclonal Antibody
Anticorps
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR). The NMDAR plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimer’s, epilepsy and ischemic neuronal cell death. Increased membrane surface expression of the NR2A subunit of the receptor has been associated synaptic plasticity.
Affinity Purified
Reconstitute in 50 μl PBS. Aliquot and store at -20°C or below. AvoidMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité