Human recombinant ACTG1 (from E. coli)
Fournisseur: ProSci Inc.
XW-RP3002
PRSIXW-RP3002EA 620 EUR InStock
PRSIXW-RP3002
Human recombinant ACTG1 (from E. coli)
Protéines et Peptides
Actins are highly conserved proteins that are involved in various types of cell motility, and maintenance of the cytoskeleton. In vertebrates, three main groups of actin isoforms, α, β and γ have been identified. The α actins are found in muscle tissues and are a major constituent of the contractile apparatus. The β and γ actins co-exist in most cell types as components of the cytoskeleton, and as mediators of internal cell motility. Actin, γ 1, encoded by this gene, is a cytoplasmic actin found in nonmuscle cells.
FUNCTION: Actins are highly conserved proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells.
SUBUNIT: Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of a two-stranded helix. Each actin can bind to 4 others.
  • This recombinant protein can be used for WB, ELISA, MS.This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité