Human recombinant PPP1CC (from E. coli)
Fournisseur: ProSci Inc.
XW-RP3205
PRSIXW-RP3205EA 773 EUR InStock
PRSIXW-RP3205
Human recombinant PPP1CC (from E. coli)
Protéines et Peptides
Protein phosphatase 1 (PP1) is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. Involved in regulation of ionic conductances and long-term synaptic plasticity. May play an important role in dephosphorylating substrates such as the postsynaptic density-associated Ca(2+)/calmodulin dependent protein kinase II.
  • This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.002% NaN, 3mM NaCl, 2.5mM
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité