TES buffer, 1X, ready to use, pH 7,8
Fournisseur: Rockland Immunochemicals
MB-058
ROCKMB-058EA 203 EUR InStock
ROCKMB-058
TES buffer, 1X, ready to use, pH 7,8
Buffers Tampons biologiques
This product is ready-to-use as a working 1X solution and requires no further dilution. This buffer consists of 10 mM Tris HCl, 100 mM NaCl, and 1,0 mM EDTA at a pH of 7,8. Meticulously prepared using ultra pure reagents dissolved in DEPC treated pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Attention: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité