ES Buffer
Fournisseur: FUJIFILM WAKO CHEMICALS
ESB-0006
WAKFESB-0006EA 360 EUR InStock
WAKFESB-0006
ES Buffer
Tampons Tampons biologiques
The ES Buffer is an endotoxin-specific buffer that is ideal for untreated LAL.
  • Each vial has solution for use in reconstituting a max of 6,0 ml of reagent
  • Ideal for use in reconstituting any LAL that has the potential to react with glucans, thus making an endotoxin-specific reagent
Though all LAL reagents produced by FUJIFILM Wako LAL are endotoxin-specific, other commercially available LAL reagents are not. Without this buffer, tests can present a potential false positive when trying to determine the endotoxin concentration in the sample.
The ES Buffer works by adding a large amount of carboxymethylated curdlan (CMC) to LAL, thus preventing the activation of LAL by glucan. The presence of large amounts of glucan does not interfere with the quantitation of endotoxin. FUJIFILM Wako first made use of these findings by developing this endotoxin-specific buffer, which contains high concentrations of CMC. When the ES Buffer is used to reconstitute LAL, reagent becomes endotoxin-specific.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité