Human recombinant TFPI2 (from cells)
Fournisseur: ProSci Inc.
PRSI91-357EA 761 EUR InStock
PRSI91-357
Human recombinant TFPI2 (from cells)
Protéines et Peptides
Human Tissue Factor Pathway Inhibitor 2 (TFPI2) has a N-terminal acidic region, three Kunita domains separated with by two linker regions, and a C-terminal basic region. TFPI2 has the function of regulating plasmin-mediated matrix remodeling, inhibits trypsin, plasmin, factor Vlla/tissue factor and weakly factor Xa.. TFPI2 has no effect on thrombin. TFPI2 may contribute to tumor progression in many cancers; in these cancers, the expression of TFPI2 can be down-regulated.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité