Human recombinant Cathepsin E (from Cells)
Fournisseur: ProSci Inc.
PRSI91-367EA 761 EUR InStock
PRSI91-367
Human recombinant Cathepsin E (from Cells)
Protéines et Peptides
Cathepsin E (CTSE) is a gastric aspartyl protease that functions as a disulfide-linked homodimer. It is a member of the Peptidase C1 family, and has a specificity similar to that of Pepsin A and Cathepsin D. CTSE is localized to the endoplasmic reticulum and Golgi apparatus, while the mature enzyme is localized to the endosome. It is expressed abundantly in the stomach, the Clara cells of the lung and activated B-lymphocytes, and at lower levels in lymph nodes, skin and spleen. CTSE is an intracellular proteinase that have a role in immune function, activation-induced lymphocyte depletion in the thymus, neuronal degeneration and glial cell activation in the brain. Futhermore, it probably involved in the processing of antigenic peptides during MHC class II-mediated antigen presentation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité