Human recombinant CD83 (from cells)
Fournisseur: ProSci Inc.
PRSI91-370EA 490 EUR InStock
PRSI91-370
Human recombinant CD83 (from cells)
Protéines et Peptides
CD83 antigen is a single-pass type I membrane protein which contains one Ig-like V-type (immunoglobulin-like) domain. CD83 is expressed by activated lymphocytes, Langerhans cells and interdigitating reticulum cells. It contains one Ig-like V-type (immunoglobulin-like) domain. , the soluble CD83 has the opposite effect and has an immune inhibitory capacity. Due to its immune inhibitory function, CD83 may play a significant role in antigen presentation or the cellular interactions that follow lymphocyte activation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité