Human recombinant C1q R1 (from Cells)
Fournisseur: ProSci Inc.
PRSI91-372EA 584 EUR InStock
PRSI91-372
Human recombinant C1q R1 (from Cells)
Protéines et Peptides
C1q R1 is also known as CD93, collectin receptor, and AA4 antigen, belongs to the Group XIV C-Type lectin family which play a role not only in cell–cell adhesion processes but also in host defence. All of them contain a C-type lectin domain, a series of epidermal growth factor like domains (EGF), a highly glycosylated mucin-like domain, a unique transmembrane domain and a short cytoplasmic tail. C1q R1 has also been identified as a hematopoietic stem cell marker.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité