Human recombinant AGER (from cells)
Fournisseur: ProSci Inc.
PRSI91-379EA 490 EUR InStock
PRSI91-379
Human recombinant AGER (from cells)
Protéines et Peptides
Advanced Glycosylation End Product-Specific Receptor (AGER) belongs to the immunoglobulin superfamily of cell surface molecules. It lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Besides AGEs, AGER is also able to bind other ligands which is thought to result in pro-inflammatory gene activation. It is known that AGER serve as a mediator of both acute and chronic vascular inflammation in certain conditions such as atherosclerosis and in particular as a complication of diabetes. Furthermore, it plays an important role in regulating the production/expression of TNF-alpha, oxidative stress, and endothelial dysfunction in type 2 diabetes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité