Human recombinant ASGPR1 (from Cells)
Fournisseur: ProSci Inc.
PRSI91-384EA 490 EUR InStock
PRSI91-384
Human recombinant ASGPR1 (from Cells)
Protéines et Peptides
Asialoglycoprotein Receptor 1 (ASGPR1) is an endocytic recycling receptor, belongs to the long-form subfamily of the C-type/Ca2+-dependent lectin family. ASGPR consists of two noncovalently-linked subnits, ASGPR1 and ASGPR2. ASGPR1 mediates the endocytosis of plasma glycoproteins, recognizes terminal galactose and N-acetylgalactosamine units. When the ligand binds to to ASGPR1, results in the complex is internalized and transported to a sorting organelle, then ASGPR1 and ligand can be disassociated, ASGPR1 returns to the cell membrane surface.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité