Human recombinant CCL14 (from Cells)
Fournisseur: ProSci Inc.
PRSI91-395EA 584 EUR InStock
PRSI91-395
Human recombinant CCL14 (from Cells)
Protéines et Peptides
Chemokine (C-C motif) Ligand 14 (CCL14) is a small cytokine belonging to the CC chemokine family. It is produced as a protein precursor that is processed to generate a mature active protein containing 74 amino acids that and is 46% identical in amino acid composition to CCL3 and CCL4. This chemokine is expressed in various tissues including spleen, bone marrow, liver, muscle, and gut. CCL14 activates monocytes, but does not induce their chemotaxis. Human CCL14 is located on chromosome 17 within a cluster of other chemokines belonging to the CC family.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité