Human recombinant CD99 (from cells)
Fournisseur: ProSci Inc.
PRSI91-403EA 490 EUR InStock
PRSI91-403
Human recombinant CD99 (from cells)
Protéines et Peptides
CD99 is a type I transmembrane glycoprotein and the founding member of the CD99 family of molecules. The extracellular domain of CD99 contains no identifiable motifs, its cytoplasmic region, although short, does have signal transduction capability. Cells known to express CD99 include fibroblasts, neutrophils, T cells, double positive thymocytes, CD34+ stem cells, monocytes and endothelial cells. Two types of CD99 isoforms have been classified. Native human CD99 is referred to as the long, or type I isoform. The best studied type II isoform shows an Asp-Gly substitution for the C terminal 27 amino acids. The type I and II isoforms have distinctive signal transduction pathways (FAKsrc for type I PI3K plus srcERK1/2 for type II), and mediate clearly different biological outcomes. Homophilic interaction between CD99 on the neutrophil and CD99 on the endothelial cell regulates the transendothelial migration of neutrophils during inflammation. Human CD99 has 48% aa sequence identity to mouse CD99.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité