Human recombinant CEACAM1 (from cells)
Fournisseur: ProSci Inc.
PRSI91-404EA 490 EUR InStock
PRSI91-404
Human recombinant CEACAM1 (from cells)
Protéines et Peptides
Carcinoembryonic Antigen-Related Cell Adhesion Molecule 1 (CEACAM1) is a member of the Carcinoembryonic Antigen (CEA) family, which belongs to the immunoglobulin superfamily. CEACAM1 is originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it is found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. CEACAM1 mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. In addition, CEACAM1 plays a important role in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité