Human recombinant CEACAM3 (from cells)
Fournisseur: ProSci Inc.
PRSI91-405EA 584 EUR InStock
PRSI91-405
Human recombinant CEACAM3 (from cells)
Protéines et Peptides
Carcinoembryonic Antigen-Related Cell Adhesion Molecule 3 (CEACAM3) belongs to the immunoglobulin superfamily and CEA family. CEACAM3 was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. CEACAM3 mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. In addition, it is associated with the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité