Human recombinant TIMP1 (from cells)
Fournisseur: ProSci Inc.
PRSI91-412EA 761 EUR InStock
PRSI91-412
Human recombinant TIMP1 (from cells)
Protéines et Peptides
Tissue Inhibitor of Metalloproteinases 1 (TIMP-1) complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Also mediates erythropoiesis in vitro; but, unlike IL-3, it is species-specific, stimulating the growth and differentiation of only human and murine erythroid progenitors. Known to act on MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-10, MMP-11, MMP-12, MMP-13, and MMP-16. TIMP-1 does not act on MMP-14.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité