Human recombinant Cystatin SN (from cells)
Fournisseur: ProSci Inc.
PRSI91-418EA 761 EUR InStock
PRSI91-418
Human recombinant Cystatin SN (from cells)
Protéines et Peptides
Cystatin-SN is a member of family 2 of the Cystatin superfamily and a cysteine proteinase inhibitor found in saliva, tears, urine, and seminal fluid. Together with Cystatins S and SA, it is produced by the salivary gland and secreted largely in the submandibular/sublingual saliva. Cystatin-SN inhibits members of the Papain family including Cathepsins B, C, H and L.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité