Human recombinant PREF-1 (from cells)
Fournisseur: ProSci Inc.
PRSI91-419EA 584 EUR InStock
PRSI91-419
Human recombinant PREF-1 (from cells)
Protéines et Peptides
Protein Delta Homolog 1 (DLK-1) is a transmembrane protein which contains a signal peptide, an extracellular domain with six tandem epidermai growth factor (EGF)-like domains, a single pass transmembrane domain, and a short cytoplasmic tail. It is found within the stromal cells in close contact to the vascular structure of placental villi, yolk sac, fetal liver, adrenal cortex and pancreas and in the beta cells of the islets of Langerhans in the adult pancreas. In addition, it is detected in some forms of neuroendocrine lung tumor tissue. DLK-1 may have a improtant role in neuroendocrine differentiation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité