Human recombinant Kallikrein 1 (from cells)
Fournisseur: ProSci Inc.
PRSI91-435EA 789 EUR InStock
PRSI91-435
Human recombinant Kallikrein 1 (from cells)
Protéines et Peptides
Kallikrein-1 (KLK1) is a member of human tissue Kallikrein family. Human KLK1 precursor contains a singal peptide (residues 1 to 18), a short pro peptide (residues 19 to 24) and a mature chain (residues 25 to 262). The function of KLK1 is to cleave Kininogen in order to release the vasoactive Kinin peptide (Lysyl-Bradykinin or Bradykinin). The Kinin peptide controls blood pressure reduction, vasodilation, smooth muscle relaxation and contraction, pain induction and inflammation. KLK1 also plays a role in angiogensis and tumorigenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité