Human recombinant MBL2 (from cells)
Fournisseur: ProSci Inc.
PRSI91-441EA 490 EUR InStock
PRSI91-441
Human recombinant MBL2 (from cells)
Protéines et Peptides
Mannose-Binding Protein C (MBP-C) belongs to the Collectin family of innate immune defense proteins. MBL binds to an array of carbohydrate patterns on pathogen surfaces. Collectin family members share common structural features: a cysteine rich amino-terminal domain, a collagen-like region, an alpha-helical coiled-coil neck domain and a carboxy terminal C-type Lectin or carbohydrate recognition domain (CRD). MBL homotrimerizes to form a structural unit joined by N-terminal disulfide bridges. These homotrimers further associates into oligomeric structures of up to 6 units. Whereas two forms of MBL proteins exist in rodents and other animals. Human MBL-2 is 25 kDa. Human MBL-2 is a secreted glycoprotein that is synthesized as a 248 amino acid (aa) precursor that contains a 20 aa signal sequence, a 21 aa cysteine-rich region, a 58 aa collagen-like segment and a 111 aa C-type lectin domain that binds to neutral bacterial carbohydrates.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité