Human recombinant neuroplastin (from cells)
Fournisseur: ProSci Inc.
PRSI91-446EA 584 EUR InStock
PRSI91-446
Human recombinant neuroplastin (from cells)
Protéines et Peptides
Neuroplastin is a 52-57 kDa member of the Ig-superfamily. Neuroplastin likely serves as a cell adhesion molecule, and is widely expressed in multiple tissues. Human Neuroplastin is 282 amino acids in length. Human Neuroplastin is a type I transmembrane glycoprotein that contions two Ig-like domains (aa 32-119 and 122-213) and a 38 aa cytoplasmic region (aa 245-282).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité