Human recombinant neurexophilin-1 (from cells)
Fournisseur: ProSci Inc.
PRSI91-447EA 525 EUR InStock
PRSI91-447
Human recombinant neurexophilin-1 (from cells)
Protéines et Peptides
Neurexophilin-1 (NXPH1) is a member of Neurexophilin family. NXPH1 consist of 271 amino acis. It contains a 21 amino acid signal peptide, 86 amino acid propeptide, and 164 amino acid mature protein. NXPH1 is expressed in subpopulations of neurons within the cerebral cortex, cerebellum and olfactory bulb that are thought to be inhibitory interneurons. In humans, NXPH2 and NXPH3 are most similar to NXPH1, sharing 84% and 64% aa identity within the mature region, respectively. By contrast, NXPH4 dost not bind a-neurexins. Genetic deletion of NXPH1 or NXPH3 produces no anatomical effect.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité