Human recombinant Tie-1 (from cells)
Fournisseur: ProSci Inc.
PRSI91-454EA 336 EUR InStock
PRSI91-454
Human recombinant Tie-1 (from cells)
Protéines et Peptides
TIE-1 (Tyrosine Kinase with Ig and EGF Homology domains 1) and TIE-2/Tek comprise a receptor tyrosine kinase (RTK) subfamily. These receptors are expressed on endothelial and hematopoietic progenitor cells and play critical roles in angiogenesis, vasculogenesis and hematopoiesis. Human TIE-1 cDNA encodes a 1124 amino acid (aa) residue precursor protein with an 18aa signal peptide, a 727 aa extracellular domain and a 354 aa cytoplasmic domain. so far, two ligands have been described for TIE-2 [angiopoietin-1 (Ang1) and angiopoietin-2 (Ang2)], but no ligand was found for TIE-1.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité