Human recombinant Chymotrypsin C (from cells)
Fournisseur: ProSci Inc.
PRSI91-469EA 789 EUR InStock
PRSI91-469
Human recombinant Chymotrypsin C (from cells)
Protéines et Peptides
Chymotrypsin C (CTRC) is a member of the peptidase S1 family. CTRC is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. CTRC has broad substrate specificity, but prefers tocleave on the carboxyl side of hydrophobic residues. CTRC is expressed primarily in the pancreas, and is secreted into the digestive tract. CTRC plays a protective role in the pancreas by mitigating premature trypsinogen activation through degradation. It has been proposed that CTRC is a key regulator of digestive zymogen activation and is a physiological coactivator of digestive carboxypeptidases proCPA1 and proCPA2. The mutation of CTRC gene encodes the digestive enzyme CTRC contribute to the development of pancreatitis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité