Human recombinant PDILT (from cells)
Fournisseur: ProSci Inc.
PRSI91-477EA 584 EUR InStock
PRSI91-477
Human recombinant PDILT (from cells)
Protéines et Peptides
Protein Disulfide-Isomerase-Like Protein of the Testis (PDILT) is a protein that belongs to the protein disulfide isomerase family. Human PDILT is synthesized as a 584 amino acid precursor that contains an 20 amino acid signal sequence and a 564 amino acid mature chain. PDILT contains 1 thioredoxin domain lacks the conserved redox-active Cys at position 417 which is replaced by a Ser residue, suggesting that it lacks thioredoxin activity. PDILT is an enzyme in the endoplasmic reticulum in eukaryotes. It is not a disulfide-linked homodimer. The PDILT protein can interacts with ERO1L and CLGN. PDILT probable redox-inactive chaperone involved in spermatogenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité