Cell Counting
Featured Solutions
EVE™ is a benchtop automatic cell counter designed to measure cell count and viability accurately and precisely. It uses the standard trypan blue technique
Learn more about EVE™
    Additional Resources
    EVE™ PLUS is a compact image-based automated cell counter that uses disposable slides to most accurately determine eukaryotic cell titers in less than 1 second. 
    Learn more about EVE™ Plus
      Additional Resources
      ADAM-MC2 is an automated fluorescence cell counter that performs cell counting and viability measurements at highest accuracy capable of replacing complex flow cytometry workflows. Combined with the disposable micro-fluidic Accuchip, the operation is extremely simple and cost-effective.
      Learn more about ADAM-MC2
        Additional Resources
        C-Slide Cell counting chamber slides for use with automated cell counter (Countess and Countess II).
        Learn more about C-Slide