![]() Nunc™ plates for cell-based assays
Product selection![]() Low attachment surface for Spheroid culture. Nunclon Sphera for spheroid-organoid cultureEnables formation of reproducible spheroid cultures. Cells grow and aggregate with virtually no cell attachment to the culture vessel. For spheroid culture, organoid culture, and 3D culture. Product information![]() Non-treated surface for suspension cultures or general assaysHigh quality, optical clear virgin polystyrene with hydrofobic surface, ideal for suspension cell culture, also useful for a variety of biochemical binding assays. ![]() Cell culture treated surface Nunclon Delta for adherent cellsA standard tissue culture (TC) surface modification that makes the polyestyrene surface more hydrofilic, thus facilitaiting maximum adhesion for a broad range of cell types. ![]() Temperature responsive surface. Nunclon Upcell for adherent culture plus trypsin-free cell harvestingEnables harvesting of cells in single cell suspension or as contiguous cell sheet by temperature reduction and can create 3D tissue models without foreign-to-the-body scaffold material. Product information![]() Standard coatings. Poly D Lysine, Colagen I for primary and finicky cellsECM-coated surface mimicking the growth environment inside the body, ideal for cells that have difficulties growing on the regular tissue culture surface. Collagen I is of animal origin, whereas PDL is fully siynthetic. Product informationMore Nunc cell based assays plates available, please check the Nunc Plates Selector Guide Literature downloads |