Nucleic Acid Purification
Featured Solutions
Omega Bio-tek Inc. offers high quality, convenient and cost-effective products for nucleic acid purification.
Learn more
cfDNA kits
    Additional Resources
    Sera-Xtracta DNA kits based on magnetic beads: Sera-Xtracta HMW DNA Kit for high yield extraction of high-molecular-weight genomic DNA from blood, buffy coat, saliva, cultured mammalian cells and solid tissue. Sera-Xtracta Cell-Free DNA Kit for efficient extraction and purification of cfDNA from plasma.
    cfDNA kit
      HMW genomic DNA kit
      peqGOLD Nucleic Acid Purification kits. High quality silica based spin column kits for a wide range of sample types. Offering High yield, high purity and value for money
      Plasmid Purification
      gDNA purification
      RNA purification