PCR Sample Protection![]() The VWR® PCR Workstation Pro HEPA offers twin decontamination action by UV inactivation of airborne and surface-bound contaminants and, therefore, represents an ideal environment for PCR sample preparation and other sensitive protocols Additional Resources |