Masterflex Ismatec Peristaltic Pump Selection Guide
Contents
Why Select Masterflex Ismatec?
Selecting a Masterflex Ismatec Pump
Selecting Ismatec Pump Tubing
Pump System and Drive Selection by Flow Range
Why Select Masterflex Ismatec?
  • Flow rates as low as 0.001 mL/min
  • High-accuracy numerical, digital, or programmable speed control
  • Tubing is the only wetted part
  • The quality of Swiss manufacturing
  • Most drives have a 2-year warranty
  • ISO 9000 Registered Supplier
  • CE compliant
Masterflex Ismatec peristaltic pumps are available as complete systems or as separate drive and pump head components. A pump system includes a drive with a permanently attached pump head; a drive alone requires a separate pump head.
Remote control capabilities make Masterflex Ismatec pumps ideal for lab automation. Most Masterflex Ismatec drives offer remote on/off, speed, and directional control. Computer interfacing is possible with all digital drives and the New Reglo pump system offers WiFi connectivity and cloud-based pump control.
The Ismatec IPC and IP range of pump systems feature a A planetary drive design in which a central gear drives all rollers results in increased flow accuracy and tubing life. Rollers turn in unison to reduce stretching and slippage of tubing.
Selecting a Masterflex Ismatec Pump
Flow Range depends on drive speed, number of rollers on the pump head, and the size of tubing. For a higher flow range, choose a pump head with fewer rollers. To reduce fluid pulsation, select a pump head with more rollers.
The number of channels refers to the number of lengths pieces of tubing that can be used simultaneously. For example, a four-channel pump head can accommodate four pieces of tubing or four cartridges. Load the four channels with different types and or sizes of tubing then pump a variety of fluids or the same fluid at different flow rates.
Speed. Fixed-speed pumps and drives are simple and easy to use, yet they accept a wide variety of tubing sizes to deliver various flow rates.
Variable-speed pumps and drives can accept analog input for remote on/off, speed, and directional control and some are controlled via cloud based app using MasterflexLive®. They are available with numerical or digital display. Digital models provide an analog output to let you monitor flow rate and feature an RS-232 interface.
Selecting Ismatec Pump Tubing
Look for the tubing information icons on each product page to determine at a glance what type of tubing to order for your pumps and pump heads. Flow rates will depend on both the rpm of your drive or pump and on the tubing diameter.
Pump System and Drive Selection by Flow Range
Model number
Flow range (mL/min)
Speed (rpm)
Channels
Rollers
Tubing type
Masterflex® Ismatec® Reglo ICC pumps
MFLX78001-43
0.001 to 35
35
2
8
3-stop
MFLX78001-45
0.001 to 35
35
8
3
3-Stop
MFLX78001-47
0.001 to 35
35
8
4
3-Stop
MFLX78001-49
0.001 to 35
24
12
4
3-Stop
MFLX78001-58
0.001 to 35
43
6
2
3-Stop
MFLX78001-60
0.001 to 35
35
8
2
3-Stop
MFLX78001-62
0.001 to 35
24
12
2
3-Stop
MFLX78001-68
0.001 to 35
43
6
3
3-Stop
MFLX78001-70
0.001 to 35
35
8
3
3-Stop
MFLX78001-72
0.001 to 35
24
12
3
3-Stop
MFLX78001-78
0.001 to 35
43
6
4
3-Stop
MFLX78001-80
0.001 to 35
35
8
4
3-Stop
MFLX78001-82
0.001 to 35
24
12
4
3-Stop
Masterflex Ismatec Reglo IPC pumps w/ adjustable occlusion cartridges
MFLX78000-03
0.002 to 44
44
8
4
2-Stop
MFLX78000-09
0.002 to 44
44
8
4
2-Stop
MFLX78000-13
0.002 to 44
44
8
8
2-Stop
MFLX78000-19
0.002 to 44
44
8
8
2-Stop
MFLX78000-23
0.002 to 44
44
8
12
2-Stop
MFLX78000-29
0.002 to 44
44
8
12
2-Stop
MFLX78000-33
0.002 to 44
44
8
16
2-Stop
MFLX78000-39
0.002 to 44
44
8
16
2-Stop
MFLX78000-43
0.002 to 44
44
8
24
2-Stop
MFLX78000-49
0.002 to 44
44
8
24
2-Stop
Masterflex Ismatec Reglo IPC Pump w/ Click-N'-Go® cartridges
MFLX78001-02
0.002 to 44
44
8
4
2-Stop
MFLX78001-08
0.002 to 44
44
8
4
2-Stop
MFLX78001-12
0.002 to 44
44
8
8
2-Stop
MFLX78001-18
0.002 to 44
44
8
8
2-Stop
MFLX78001-22
0.002 to 44
44
8
12
2-Stop
MFLX78001-28
0.002 to 44
44
8
12
2-Stop
MFLX78001-32
0.002 to 44
44
8
16
2-Stop
MFLX78001-38
0.002 to 44
44
8
16
2-Stop
MFLX78001-42
0.002 to 44
44
8
24
2-Stop
MFLX78001-48
0.002 to 44
44
8
24
2-Stop
Masterflex Ismatec Reglo IPC-N pumps w/ adjustable occlusion cartridges
MFLX78000-11
0.0004 to 11
11
8
8
2-Stop
MFLX78000-17
0.0004 to 11
11
8
8
2-Stop
MFLX78000-31
0.0004 to 11
11
8
16
2-Stop
MFLX78000-37
0.0004 to 11
11
8
16
2-Stop
MFLX78000-41
0.0004 to 11
11
8
24
2-Stop
MFLX78000-47
0.0004 to 11
11
8
24
2-Stop
Masterflex Ismatec Reglo IPC-N pumps w/ Click-N'-Go cartridges
MFLX78001-00
0.0004 to 11
11
8
4
2-Stop
MFLX78001-06
0.0004 to 11
11
8
4
2-Stop
MFLX78001-10
0.0004 to 11
11
8
8
2-Stop
MFLX78001-16
0.0004 to 11
11
8
8
2-Stop
MFLX78001-20
0.0004 to 11
11
8
12
2-Stop
MFLX78001-26
0.0004 to 11
11
8
12
2-Stop
MFLX78001-30
0.0004 to 11
11
8
16
2-Stop
MFLX78001-36
0.0004 to 11
11
8
16
2-Stop
MFLX78001-40
0.0004 to 11
11
8
24
2-Stop
MFLX78001-46
0.0004 to 11
11
8
24
2-Stop
Masterflex Ismatec Reglo IP pumps w/ adjustable occlusion cartridges
MFLX78010-03
0.002 to 44
44
8
4
2-Stop
MFLX78010-09
0.002 to 44
44
8
4
2-Stop
MFLX78010-13
0.002 to 44
44
8
8
2-Stop
MFLX78010-19
0.002 to 44
44
8
8
2-Stop
MFLX78010-23
0.002 to 44
44
8
12
2-Stop
MFLX78010-29
0.002 to 44
44
8
12
2-Stop
MFLX78010-33
0.002 to 44
44
8
16
2-Stop
MFLX78010-39
0.002 to 44
44
8
16
2-Stop
MFLX78010-43
0.002 to 44
44
8
24
2-Stop
MFLX78010-49
0.002 to 44
44
8
24
2-Stop
MFLX78010-53
0.002 to 44
44
8
24
2-Stop
MFLX78010-59
0.002 to 44
44
8
24
2-Stop
Masterflex Ismatec Reglo IP pumps w/ Click-N'-Go cartridges
MFLX78011-03
0.002 to 44
44
4
4
2-Stop
MFLX78011-09
0.002 to 44
44
4
4
2-Stop
MFLX78011-13
0.002 to 44
44
4
8
2-Stop
MFLX78011-19
0.002 to 44
44
4
8
2-Stop
MFLX78011-23
0.002 to 44
44
4
12
2-Stop
MFLX78011-29
0.002 to 44
44
4
12
2-Stop
MFLX78011-33
0.002 to 44
44
4
16
2-Stop
MFLX78011-39
0.002 to 44
44
4
16
2-Stop
MFLX78011-43
0.002 to 44
44
4
24
2-Stop
MFLX78011-49
0.002 to 44
44
4
24
2-Stop
Masterflex Ismatec Reglo IP-N pumps w/ adjustable occlusion cartridges
MFLX78010-11
0.0004 to 11
11
8
8
2-Stop
MFLX78010-17
0.0004 to 11
11
8
8
2-Stop
MFLX78010-27
0.0004 to 11
11
8
12
2-Stop
MFLX78010-41
0.0004 to 11
11
8
24
2-Stop
MFLX78010-47
0.0004 to 11
11
8
24
2-Stop
Masterflex Ismatec Reglo IP-N pumps w/ Click-N'-Go cartridges
MFLX78011-01
0.0004 to 11
11
4
4
2-Stop
MFLX78011-07
0.0004 to 11
11
4
4
2-Stop
MFLX78011-11
0.0004 to 11
11
4
8
2-Stop
MFLX78011-17
0.0004 to 11
11
4
8
2-Stop
MFLX78011-21
0.0004 to 11
11
4
12
2-Stop
MFLX78011-27
0.0004 to 11
11
4
12
2-Stop
MFLX78011-41
0.0004 to 11
11
4
24
2-Stop
MFLX78011-47
0.0004 to 11
11
4
24
2-Stop
Masterflex Ismatec MiniFlex® single-channel pump
MFLX78018-40
0.004 to 365
350
1
N/A
L/S 13, 14, 16, 25
Masterflex Ismatec Miniflex dual-channel pump
MFLX78018-42
0.004 to 190
350
1
N/A
L/S 13, 14, 16