Viral Clearance breakthrough: Enhancing efficacy and protein stability with a regulatory compliant, novel detergent
Webinar
On demand
Replay
Implementing of effective viral clearance processes during biologics manufacturing are the final shield to ensure product and patient safety. One of the classical methods for viral inactivation (VI) is the detergent treatment step in manufacturing of various drug modalities, including antibodies, plasma-derived products and AAV. In this presentation, we will discuss on the application of a novel biodegradable detergent to enhance VI while in compliance with regulation. An extensive study with different model viruses demonstrated its capability of providing >6.5 Log of viral inactivation and high virus killing kinetics independent of operational temperatures (4 ⁰C to RT). Other benefits of using this detergent include no environmental toxicity, no impact on product stability or process performance. This detergent can easily be removed from the product and be detected by a simple analytical assay.
This presentation was given at BPI Europe, April 11, 2024.