J.T.Baker® Viral Inactivation Solution
Viral Inactivation with high purity and quality
Improve viral inactivation efficiency and minimize introduction of protein and process related impurities with J.T.Baker® Viral Inactivation Solution. This biodegradable, ready-to-use detergent maintains cGMP quality and regulatory compliance while addressing environmental concerns with REACH.
Maximize downstream efficiency and minimize risk
The new readily biodegradable J.T.Baker® Viral Inactivation Solution from Avantor® is a highly effective detergent that efficiently inactivates or destroys residual viruses prior to the Protein A capture step. Compatible across a range of molecules, J.T.Baker® Viral Inactivation Solution minimizes contamination risk by reducing the viral burden on subsequent process steps without impacting protein integrity.
Superior efficiency & performance
High purity reduces downstream risk
Ease of use offers resource savings
J.T.Baker® Viral Inactivation Solution
Seeking a sustainable solution for viral inactivation?
Choose the safer choice for improved efficiency, compatibility, and regulatory compliance while being mindful of environmental considerations.
Maximize efficiency in your viral clearance process in downstream.
    Related content