Wilt u opnieuw aanmelden?
Avantor's online ervaring verandert om u beter van dienst te kunnen zijn! Vanaf 4 april 2025 worden klanten overgezet naar een nieuw platform om de digitale winkelervaring te optimaliseren.
Anti-CCT2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverancier: Bioss
BS-11267R-A350
BOSSBS-11267R-A350EA 893 EUR InStock
BOSSBS-11267R-A350
Anti-CCT2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
CCT2 is one of eight largely unrelated subunit proteins found in a protein chaperone complex known as the chaperonin-containing TCP-1 (CCT) or TRiC complex. The CCT complex is an abundanct cytoslic component that is credited with helping newly synthesized polypeptides adopt the correct conformation (1). Proteins that fold and assemble with the help of CCT include the cytoskeletal proteins actin and tubulin as well as up to 15% of newly synthesized eukaryotic proteins (2). CCT2 is the β-subunit of the chaperone complex and is one of several CCT proteins that exhibit increased expression inMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité