Wilt u opnieuw aanmelden?
Avantor's online ervaring verandert om u beter van dienst te kunnen zijn! Vanaf 4 april 2025 worden klanten overgezet naar een nieuw platform om de digitale winkelervaring te optimaliseren.
Anti-APOC4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverancier: Bioss
BS-6338R-A488
BOSSBS-6338R-A488EA 893 EUR InStock
BOSSBS-6338R-A488
Anti-APOC4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antilichamen
This gene encodes a lipid-binding protein belonging to the apolipoprotein gene family. The protein is thought to play a role in lipid metabolism. Polymorphisms in this gene may influence circulating lipid levels and may be associated with coronary artery disease risk. This gene is present in a cluster with other related apolipoprotein genes on chromosome 19. Naturally occurring read-through transcription exists between this gene and the neighboring downstream apolipoprotein C-II (APOC2) gene.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOC4
Clonality: Polyclonal
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité