IDS gate
Fournisseur: WTW
1FD680
WTWA1FD680EA 1330 EUR InStock
WTWA1FD680
IDS gate
Enregistreurs de données
Gateway with REST API interface for the allocation of data records. Suitable for pH, ORP, conductivity and dissolved oxygen of IDS wireless sensors.
  • Safe, automated measurement data collection
  • Manipulation proof, GxP compliant additional data (meta data)
  • Future proof data format (JSON)
The IDS Gate is a software interface (API) that can be used to integrate IDS wireless sensors into an existing network. Via the interface, measured data and meta data can be automatically transferred to a database system (e.g. LIMS) or a spreadsheet program (e.g. Microsoft Excel). Data is transmitted wirelessly via a barcode reader or the IDS wireless sensors. The IDS Gate provides the data in JSON format. JSON is a data format that ensures that the data can be read very easily both by machine evaluation systems, such as LIMS or Excel, and by humans. The data transfer is encrypted via HTTPS. For digital long-term archiving, the data can also be provided as PDF/A in addition to JSON. Browser-based visualisation makes it possible to display data in real time on networked hardware such as smartphones, tablets or monitors, independent of operating systems.
Information de livraison: Includes power supply and antenna for Wi-Fi/BT.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité