15005.2000
15000.2000
15025.2000
15030.2000
15035.2000
15095.2000
15100.2000
15105.2000
15110.2000
15115.1000
18693.2000
17746.2000
21085.2000
21084.2000
21083.2000
21082.2000
21081.2000
17745.2000
18453.2000
18452.2000
21080.2000
21079.2000
15650.2000
15439.2000
21078.2000
KRAF15005.2000EA
175
EUR
InStock
KRAF15005.2000
KRAF15000.2000
KRAF15025.2000
KRAF15030.2000
KRAF15035.2000
KRAF15095.2000
KRAF15100.2000
KRAF15105.2000
KRAF15110.2000
KRAF15115.1000
KRAF18693.2000
KRAF17746.2000
KRAF21085.2000
KRAF21084.2000
KRAF21083.2000
KRAF21082.2000
KRAF21081.2000
KRAF17745.2000
KRAF18453.2000
KRAF18452.2000
KRAF21080.2000
KRAF21079.2000
KRAF15650.2000
KRAF15439.2000
KRAF21078.2000
Conductiviteit standaard (25°C; 0,01 mol/l; KCl)
Standaarden
Geleidbaarheidsstandaarden
A complete range of certified control standards with values ranging from 1,3 to 500 000 µS/cm to meet all the requirements for electrochemical analysis.
- Maximum deviation ±1%
- Standardised for 25 °C
Certificaten: NIST certified.