985033
985028
963026
985023
985022
985012
985030
985011
985026
985027
985029
963029
985036
985038
985020
MANA985033EA
85.7
EUR
InStock
MANA985033
MANA985028
MANA963026
MANA985023
MANA985022
MANA985012
MANA985030
MANA985011
MANA985026
MANA985027
MANA985029
MANA963029
MANA985036
MANA985038
MANA985020
NANOCOLOR® Photometric Tube Tests, COD
Milieutestkits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of the COD (Chemical oxygen demand) in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes
The tests are particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.