Synoniemen:
Caustic soda
38227-1EA
38227-6X1EA
38227-1EAEA
61.8
EUR
InStock
38227-1EA
38227-6X1EA
Natriumhydroxide concentrate for dilution to 1 L, 0.01 M NaOH (0.01 N) volumetrische oplossing, Fluka™
Natriumhydroxide
Concentrate for 1 L standard solution