Synoniemen:
Methoxyacetic methyl ester
B22528.14
B22528.22
B22528.36
B22528.14EA
28.8
EUR
InStock
B22528.14
B22528.22
B22528.36
Methyl methoxyacetate ≥99%
Methyl methoxyacetate
Purity: 99%
CAS No.: 6290-49-9
Molecular Formula: CH3OCH2CO2CH3
Formula Weight: 104.11
Flash Point: 35°(95°F)
Refractive Index: 1.3960
Boiling Point: 129-130°
Beilstein Reference: 1742944
UN No.: 3272
MDL No.: MFCD00008451