Blijf aangemeld
Verloopt binnen 
De sessie is verlopen.
Uw sessie is verlopen. Voor uw veiligheid bent u afgemeld.
Wilt u opnieuw aanmelden?
Masterflex® I/P® MasterSense™ Process Drives
Reimage bioprocessing
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Shop Drives
Shop Complete Systems
Take your process to the next level
1
Perfect for sanitary environments-
316 polished stainless steel housing
IP66 NEMA 4X rated for washdown
2
Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
3
Intuitive digital touchscreen with optimized menu navigation to store and recall standard protocols ensuring continuity - even with wet gloves on!
4
Over-the-air firmware updates ensure optimization of your pump system without moving the system while MasterflexLive® cloud storage system securely stores, monitors and generates reports for all Masterflex pump systems
VIEW FULL SPECS
Masterflex L/S MasterSense Peristaltic Stainless Steel Washdown Process Pump Drive Features
5
Powerful performance with a broad flow range- 0.0006 to 26 LPM. Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
OTHER DRIVE OPTIONS
6
Convenient open-head sensor and antidrip features. Accepts all I/P pump heads.
SHOP I/P PUMP HEADS
7
Graphical indicator validates optimal tube calibration. Choose from over 20 formulations and sizes of pump tubing.
SHOP I/P PUMP TUBING
DOWNLOAD FLYER
Programmable Continuous Run Pumping
Continuous Mode
Programmable Timed Dispensing
Time Mode
Programmable Volume Pumping
Volume Dispense
Pressure Sensor
Pressure Sensor
User Access
User Access
Program Save & Recall
Program Save & Recall
Network Detection
Network Detection
remote monitor
Remote Mode
Optimal Tubing Load Indicator
Tube Calibration
Set up is a breeze
Intuitive touch screen interface, even wearing gloves
  • Easy to set up and program
  • 7" capacitive TFT/RGB touch screen with a clear, wide viewing angle
  • Adjust parameters with a simple touch-even with gloves on
  • Operational modes include continuous run, volume dispense, and time dispense
  • Batch control and interval/off time in Volume and Time modes
  • Antidrip and speed ramping available in Volume mode-ensure accuracy and minimize sample loss
  • Easily save and recall up to 50 commonly used protocols, each with a unique name
  • Know when you hit your mark-graphical indicator lets you know when loaded tubing is optimally calibrated
MasterSense monitors your media so you don't have to
With the extra time, a process engineer could:
Advanced Connectivity and Control Options
Monitor from anywhere
MasterflexLive Remote Pump Monitoring
MasterflexLive
  • MasterflexLive is our secure, cloud-enabled platform for real-time remote monitoring of all Masterflex pump parameters
  • Monitor critical processes running 24/7 without the need for onsite personnel
  • Push notifications alert you to changes in operating conditions and error messages
Learn more
Flexible Connectivity
  • Your choice of connectivity options-MasterflexLive®, EtherNet/IP, or Profibus and RS232 options
  • Three-tier access to pump functionality-use this option to protect your process by controlling who can operate the pump and at what level
  • Over-the-air firmware updates-automatically allows you to take full advantage of future enhancements
  • Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
See full specifications
NEW Masterflex® I/P® Rapid-Load Pump Head
Built for bioprocess applications - designed for ease of use and process flexibility
Learn more
Easy Integration and Scale-up
Flow Rate up to 42 LPM
Minimize risk with a closed system. Non-contaminating, low shear, and noninvasive makes this pump perfect for Biopharma and healthcare.
Flow Rate up to 42 LPM
Three-tier access and the ability to save and recall programs make your fluidic process secure and accurate.
Flow Rate up to 42 LPM
The I/P series is ideal for mimicking large-scale (up or out) systems, which allows quicker, consistent process optimization.
Precision manufactured pump tubing ensures accuracy, reliability, fluid compatibility and the regulatory approvals needed for critical process applications.
Complete Specifications & Product Details
Types of pump heads accepted 0.1 to 650 RPM (up to 2 pump heads accepted) 0.1 to 650 RPM Complete System with Easy-Load 77602-30 and C-Flex ULTRA® tubing 06434-73
Flow Range 0.0006 to 19 LPM 0.001 to 8.0 LPM
With EtherNet/IP and MasterflexLive MFLX77421-00 MFLX77966-00
With Profibus and MasterflexLive MFLX77421-02 MFLX77966-10
Drive Specifications
Speed control accuracy (resolution) ±0.1% (0.1 rpm at 650 rpm)
Display 7” capacitive TFT/RGB touch screen with in-plane switching (wide viewing angle); 800 x 480 pixels
External control - Input 0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Start/Stop, Direction (CW/CCW), Prime via contact closure
External Control - Output 0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Tach output (open collector), Motor running, Open head, Local/Remote, General alarm
Motor BLDC with encoder, continuous duty, reversible, 250 W (0.34 hp)
Open head sensor Yes; extension cable included
Housing materials 316 stainless steel housing and bezel with anodized aluminum base
Enclosure rating IP66, NEMA 4X
Agency approvals UL, cUL, CE
Warranty 2 years
Dimensions (L x W x H) 14" x 10.2" x 13.6" (35.6 x 26.0 x 34.6 cm)
Shipping weight 19.5 kg (43 lb)
Power 90 to 260 VAC, 50/60 Hz
I/P Pump Tubing Flow Ranges in LPM
rpm I/P Precision pump tubing I/P High-performance Precision pump tubing
I/P 26 I/P 73 I/P 82 I/P 70 I/P 88 I/P 89
0.1 to 650 0.0006 to 4.0 0.001 to 8.0 0.002 to 13 0.001 to 8.0 0.002 to 17 0.002 to 19