Antibiotics & Antimycotics
Antibiotics and Antimycotics
Antibiotics and Antimycotics are a type of antimicrobial drug used in the treatment and prevention of bacterial infections. They may either kill or inhibit the growth of microorganisms.
In science those substances are mostly used in cell culture. They are used to select for a specific type of cells. Either when cultivating mammal cells to exclude bacteria or e.g. to select for genetically modified cells which carry a resistance gene for selection on them.
PanReac AppliChem antibiotics and antimycotics prove to be used from small assays to bigger incubators. On this page you can find a selection of our finest most common used products from this product range.
Also here it is about trust and reproducibility for all your experiments and growing in cell culture or microbiology.
List of selected antibiotics:
Product No. Description Target
organism
Mode
of action
Recommended
working
concentration
Stock
solution
A1907
Amphotericin B
fungi, yeast
binds to sterols with planar structure and impairs with membrane permeability
0.25 µg/ml
>3 µg/ml fungicidal
30-40 mg/ml in DMSO
A3784
Blasticidin S hydrochloride
Prokaryotes, Eukaryotes
inhibits protein biosynthesis by preventing the formation of the peptide bond
3-100 µg/ml
50 mg/ml in H2O or buffer store at +4°C - -20°C
A1491
Carbenicillin disodium salt
gram negative germs, enterococci
inhibits cell wall synthesis (transpeptidase) in growing bacteria
20-60 µg/ml
50 mg/ml in H2O/MeOH 1:1 store at -20°C
A0879
Cycloheximide (Actidion)
fungi, eukaryotes
binds to 80 S ribosome in eukaryotic cells; inhibits formation of peptide bond
10 µg/ml
10 mg/ml
A1492
Gentamycin sulfate
gram positive/ negative germs
inhibits protein synthesis by binding to the L6 protein of the 50 S ribosomal subunit
15-50 µg/ml
10-20 mg/ml in water, formamide
A2175
Hygromycin B - Solution
10-400 µg/ml
ca. 41 mg/ml in H2O; store at -20°C
A1493 & A4789
Kanamycin sulfate & Kanamycin Sulfate (Ph. Eur., BP) pure, pharma rade
gram positive/
negative cocci & bacteria
inhibits protein synthesis (translocation)
10-100 µg/ml
10 mg/ml in H2O
store at -20°C
A0890
Polymyxin B sulfate
gram negative bacteria - non- proliferating germs too
interaction with phospholipid components of the bact. cell membrane; changes permeability of the membrane and causes efflux of essential plasma compounds
50 µg/ml
25 mg/ml H2O, MeOH
A1839
Vancomycin hydrochloride
bacteriostatic and bacterizide against gram positive cocci and bacteria
amphoteric glyopeptide antibiotic; binds to bacterial cell wall precursors (peptidoglycans)
1-25 µg/ml
soluble in water ›100 mg/ml
Request Quote