Automated Nucleic Acid Purification at your Fingertips from Small to Large Volume Samples
Webinar
Thursday, 13th February, 2025  
15:00-16:00 CET (Berlin, Paris, Madrid)
Nucleic acid purification is the first crucial step in genomics downstream applications such as sequencing, PCR, transfection and more. However, traditional spin-column-based DNA and RNA extraction workflows are time-consuming. What if there is a new way to free researchers and let them focus on other priorities? Join this webinar to discover more about the new standards in high and low-volume nucleic acid purification.
You will learn from an Omega Bio-tek expert how the new MagBinder® Fit24 can become your ultimate automated solution for reliable nucleic acid extraction from a wide range of sample types.
The MagBinder® Fit24 introduces a convenient and efficient method for extracting high-quality DNA and RNA from up to 4 mL of sample volume, while still eluting in 50 µL.
With the magnetic bead basis of this technology, the nucleic acid extraction process is automated post-lysis. Less hands-on work, more reliable results — sign up to learn more!
For more information, please contact us at webinar@avantorsciences.com
Watch the replay
Presented By:
Brandon_Easparro   
Brandon Easparro
Brandon Easparro oversees the life cycle of products from design to market success. He spent time as a lab manager at Omni International, Inc. before joining Omega Bio-tek as a product support scientist, ultimately transitioning to his role as a Product Manager. Lab experience and in-depth product knowledge allow Brandon to clearly understand what it takes to consistently deliver high quality products that meet customers’ needs.