NORPRENE® (A 60 G) Tubing
Advantages:
  • Best choice for vacuum/pressure applications.
  • Offers longest life with good flow consistency.
  • Heat and ambient ozone resistant.
  • Good resistance to acids and alkalies.
  • Black color hides dirt and dust.
  • Heat sealable, nonaging, and nonoxidizing.
  • High dielectric constant.
Limitations:
  • Potential leaching of USP mineral oil or blend material.
Application Suitability:
Acids: Good
Alkalies: Good
Organic solvents: Not recommended
Pressure: Excellent
Vacuum: Excellent
Viscous fluids: Excellent
Sterile fluids: Not recommended
Physical Characteristics and Compostion:
  • Thermoplastic elastomer.
  • Polypropylene-based material with USP mineral oil.
  • Excellent tensile strength.
  • Firm (stiff) material.
  • Opaque, black.
Temperature Range:
-75 to 275°F (-59 to 135°C)
Meets Classifications:
  • None
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2: 1200
H2: -
O2: 200
N2: 80
Cleaning/Sterilization:
  • Sterilization is not recommended.