Silicone (platinum-cured) Tubing
Advantages:
  • Excellent biocompatibility; no leachable additives, DOP, or plasticizers; odorless and nontoxic, fungus resistant.
  • No taste imparted to transported fluids.
  • Extremely good at low temperatures.
  • Weather, ozone, corona, and radiation resistant.
  • Minimal tendency to take a set.
  • See additional notes in "Silicone Tubing" for differences between platinum- and peroxide-cured silicone tubing formulations.
Limitations:
  • Do not use with concentrated solvents, oils, or acids.
  • Relatively high gas permeability.
  • See additional notes in "Silicone Tubing"
Application Suitability:
Acids: Poor
Alkalies: Poor
Organic solvents: Not recommended
Pressure: Fair
Vacuum: Good
Viscous fluids: Fair
Sterile fluids: Excellent
Physical Characteristics and Composition:
  • Thermal set rubber.
  • Siloxane polymers and amorphous silica.
  • Excellent compression strength.
  • Soft material.
  • Translucent, clear to light amber.
Temperature Range:
-60 to 460°F (-51 to 232°C)
Meets Classifications:
  • USP Class VI
  • EP
  • FDA 21 CFR 177.2600
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2: 20,132
H2: 6579
O2: 7961
N2: 2763
Cleaning/Sterilization:
  • Clean with hot water/soap solution.
  • Use nonoily soap such as IVORY.
  • Do not use synthetic detergents or oil-based soaps as they may be absorbed by the tubing and subsequently leach out into the fluid.
  • Rinse thoroughly with distilled water.
  • Sterilize as described in "Silicone Tubing"
  • Ethylene oxide (ETO) sterilization is not recommended—sufficient data are not available regarding complete outgassing of residual ETO and ETO byproducts.
Last Updated:
Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.
Pour en savoir plus, consultez notre Politique de Confidentialité..