Basics of Centrifugation
Reprinted with permission of THERMO
The purpose of this tutorial is to introduce basic concepts of centrifugation, including vocabulary, centrifuge and rotor types, separation techniques, and even gradient selection. For further details regarding centrifugation, please refer to the Sorvall® and Heraeus recommended reading the literature cited in the reference section.
Contents.
I. Introduction
II. Increasing the effect of gravity: the centrifuge
III. Types of centrifugal separation
IV. Rotor categories
V. Selection of centrifuge tubes
VI. Common centrifuge vocabulary and formulas
VII. Reference and suggested readings
I. Introduction.
Centrifugation is one of the most important and widely applied research techniques in biochemistry, cellular and molecular biology, and in medicine. Current research applications rely on isolation of cells, subcellular organelles, and macromolecules, often in high yields.
A centrifuge uses centrifugal force (g-force) to isolate suspended particles from their surrounding medium on either a batch or a continuous-flow basis. Applications for centrifugation are many and may include sedimentation of cells and viruses, separation of subcellular organelles, and isolation of macromolecules such as DNA, RNA, proteins, or lipids.
II. Increasing the effect of gravity: the centrifuge.
Many particles or cells in a liquid suspension, given time, will eventually settle at the bottom of a container due to gravity (1 x g). However, the length of time required for such separations is impractical. Other particles, extremely small in size, will not separate at all in solution, unless subjected to high centrifugal force. When a suspension is rotated at a certain speed or revolutions per minute (RPM), centrifugal force causes the particles to move radially away from the axis of rotation. The force on the particles (compared to gravity) is called Relative Centrifugal Force (RCF). For example, an RCF of 500 x g indicates that the centrifugal force applied is 500 times greater than Earth’s gravitational force. Table 1 illustrates common centrifuge classes and their applications.
Table 1. Classes of centrifuges and their applications.
  Centrifuge Classes
Lowspeed High-speed Ultra/micro-ultra
Maximum Speed (rpm x 103) 10 28 100/150
Maximum RCF (x103) 7 100 800/900
Pelleting applications
Bacteria Yes Yes (Yes)
Animal and plant cells Yes Yes (Yes)
Nuclei Yes Yes (Yes)
Precipitates Some Most (Yes)
Membrane fractions Some Some Yes
Ribosomes/Polysomes - - Yes
Macromolecules - - Yes
Viruses - Most Yes
( ) = can be done but not usually used for this purpose
III. Types of Centrifugal Separations.
1. Differential centrifugation. Separation is achieved primarily based on the size of the particles in differential centrifugation. This type of separation is commonly used in simple pelleting and in obtaining partially-pure preparation of subcellular organelles and macromolecules. For the study of subcellular organelles, tissue or cells are first disrupted to release their internal contents. This crude disrupted cell mixture is referred to as a homogenate. During centrifugation of a cell homogenate, larger particles sediment faster than smaller ones and this provides the basis for obtaining crude organelle fractions by differential centrifugation. A cell homogenate can be centrifuged at a series of progressively higher g-forces and times to generate pellets of partially-purified organelles.
When a cell homogenate is centrifuged at 1000 x g for 10 minutes, unbroken cells and heavy nuclei pellet to the bottom of the tube. The supernatant can be further centrifuged at 10,000 x g for 20 minutes to pellet subcellular organelles of intermediate velocities such as mitochondria, lysosomes, and microbodies. Some of these sedimenting organelles can obtained in partial purity and are typically contaminated with other particles. Repeated washing of the pellets by resuspending in isotonic solvents and re-pelleting may result in removal of contaminants that are smaller in size (Figure 1). Obtaining partially-purified organelles by differential centrifugation serves as the preliminary step for further purification using other types of centrifugal separation (density gradient separation).
2. Density gradient centrifugation. Density gradient centrifugation is the preferred method to purify subcellular organelles and macromolecules. Density gradients can be generated by placing layer after layer of gradient media (Table 2) such as sucrose in a tube with the heaviest layer at the bottom and the lightest at the top in either a discontinuous or continuous mode. The cell fraction to be separated is placed on top of the layer and centrifuged. Density gradient separation can be classified into two categories. 2a. Rate-zonal (size) separation. 2b. Isopycnic (density) separation.
2a. Rate zonal (size) separation
Rate-zonal separation takes advantage of particle size and mass instead of particle density for sedimentation. Figure 2 illustrates a rate-zonal separation process and the criteria for successful rate-zonal separation. Examples of common applications include separation of cellular organelles such as endosomes or separation of proteins, such as antibodies. For instance, Antibody classes all have very similar densities, but different masses. Thus, separation based on mass will separate the different classes, whereas separation based on density will not be able to resolve these antibody classes.
Certain types of rotors are more applicable for this type of separation than others. Please See rotor categories (below) and Table 2.
Criteria for successful rate-zonal centrifugation:
  • Density of the sample solution must be less than that of the lowest density portion of the gradient.
  • Density of the sample particle must be greater than that of the highest density portion of the gradient.
  • The pathlength of the gradient must be sufficient for the separation to occur.
  • Time is important. If you perform too long runs, particles may all pellet at the bottom of the tube.
2b. Isopycnic separation
In this type of separation, a particle of a particular density will sink during centrifugation until a position is reached where the density of the surrounding solution is exactly the same as the density of the particle. Once this quasi-equilibrium is reached, the length of centrifugation does not have any influence on the migration of the particle. A common example for this method is separation of nucleic acids in a CsCl gradient. Figure 3 illustrates the isopycnic separation and criteria for successful separation. A variety of gradient media can be used for isopycnic separations and their biological applications are listed in Table 2.
Criteria for successful isopycnic separation:
  • Density of the sample particle must fall within the limits of the gradient dens
    ities.
  • Any gradient length is acceptable.
  • The run time must be sufficient for the particles to band at their isopycnic point. Excessive run times have no adverse effect.
Table 2. Applications of density gradient media for isopycnic separations.
Gradient media Cells Viruses Organelles Nucleoproteins Macro-molecules
Sugars
(e.g. sucrose)
+ +++ +++ + -
Polysaccharides
(e.g. Ficoll)
++ ++ ++ - -
Colloidal silica
(e.g. Percoll)
+++ + +++ - -
Iodinated media
(e.g. Nycodenz)
++++ ++ ++++ +++ +
Alkali metal salts
(e.g. CsCl)
- ++ - ++ ++++
++++ excellent, +++ good, ++ good for some applications, + limited use, - unsatisfactory
Source: D. Rickwood, T.C. Ford, J. Steensgard (1994) Centrifugation essential data, John Wiley & Sons Ltd. U.K.
IV. Rotor categories
Rotors can be broadly classified into three common categories namely swinging-bucket rotors, fixed-angle rotors, and vertical rotors (Figure 4, Table 3). Note that each type of rotor has strengths and limitations depending on the type of separation.
  1. swinging-bucket
  2. fixed-angle
  3. vertical
Other rotors include continuous flow and elutriation rotors.
Table 3. Types of rotors and their applications.
Type of rotor Pelleting Rate-zonal Sedimentation Isopycnic
Fixed-angle Excellent Limited Variable*
Swinging-Bucket Inefficient Good Good**
Vertical NS Good Excellent
Zonal NS Excellent Good
NS = not suitable
*Good for macromolecules, poor for cells, and organelles
**Good for cells and organelles, caution needed if used with CsCl
In swinging bucket rotors, the sample tubes are loaded into individual buckets that hang vertically while the rotor is at rest. When the rotor begins to rotate the buckets swing out to a horizontal position (Figure 4). This rotor is particularly useful when samples are to be resolved in density gradients. The longer pathlength permits better separation of individual particle types from a mixture. However, this rotor is relatively inefficient for pelleting. Also, care must be taken to avoid “point loads” caused by spinning CsCl or other dense gradient materials that can precipitate.
In fixed-angle rotors, the sample tubes are held fixed at the angle of the rotor cavity. When the rotor begins to rotate, the solution in the tubes reorients (Figure 4). This rotor type is most commonly used for pelleting applications. Examples include pelleting bacteria, yeast, and other mammalian cells. It is also useful for isopycnic separations of macromolecules such as nucleic acids.
In vertical rotors, sample tubes are held in vertical position during rotation. This type of rotor is not suitable for pelleting applications but is most efficient for isopycnic (density) separations due to the short pathlength. Applications include plasmid DNA, RNA, and lipoprotein isolations.
V. Selection of Centrifuge Tubes.
Table 4 and Table 5 illustrate properties of centrifuge tubes and the proper rotors in which they should be used.
Selection of the appropriate centrifuge tube:
Major factor in selection of a tube (plastic) material:
  • Prevents sample leakage or loss
  • Ensures chemical compatibility
  • Allows easy sample recovery
  • Clarity
  • Chemical resistance
  • Sealing mechanism (if needed)
    Table 4- Chemical Compatibility of Popular Tube Materials
    Tube Plastic Type Clarity Chemical Resistance*
    Polypropylene (PP) Opaque Good
    Polyallomer (PA) Opaque Good
    Polycarbonate (PC) Clear Poor
    Polyethylene terephtalate (PET) Clear Poor
    * For more information, please check out chemical resistance chart available on this website.
  • check product guide pages or tube packaging for notes on recommended sample volume and maximum speed.
  • always run thin-walled, sealed tubes full in a fixed angle or vertical rotor.
    Examples:
    - open top tube with multiple sealing assembly
    - Re-seal tubes
    - Ultracrimp® and Clearcrimp® tubes
  • autoclave tubes only if absolutely necessary and only at 121°C for 15 min.
  • avoid cleaning plastic tubes in automated dishwashers or glassware washers, which may produce excessively hot temperatures.
  • we recommend that you clean tubes with a mild laboratory detergent in warm water, rinse, and thin air dry.
  • tube must be carefully matched with rotor type to prevent sample loss and/or failure as illustrated in Table 5 below.
To prolong tube life and avoid breakage or collapse:
Table 5 - Tube Type and Rotor Compatibility
  Rotor Type
Tube type Fixed-angle Swinging-bucket Vertical
Thin wall open top No Yes No
Thick wall open top Yes Yes No
Thin wall sealed Yes Some tube types Yes
Oak ridge Yes No No
VI. Common Centrifugation Vocabulary and Formulas.
  • Pellet: hard-packed concentration of particles in a tube or rotor after centrifugation.
  • Supernatant: The clarified liquid above the pellet.
  • Adapter: A device used to fit smaller tubes or centrifugal devices in the rotor cavities.
  • RPM: Revolutions Per Minute (Speed).
  • Rmax: Maximum radius from the axis of rotation in centimeters.
  • Rmin: Minimum radius from the axis of rotation in centimeters.
  • RCF: Relative centrifugal Force. RCF = 11.17 x Rmax (RPM/1000)2
  • K-factor: Pelleting efficiency of a rotor. Smaller the K-factor, better the pelleting efficiency.
  • S-value: the sedimentation coefficient is a number that gives information about the molecular weight and shape of the particle. S-value is expressed in Svedberg units. The larger the S-value, the faster the particle separates.
    For more information about sedimentation coefficients, please refer to the section on references and suggested readings in this article.
  • Pelleting time: time taken to pellet a given particle. T = K/S where T= pellet time in hours. K = K-factor of the rotor, and S = sedimentation coefficient.
  • Rotor conversion formula: If the time to pellet a sample in your “old” rotor is known, one could determine the time it would take for the same sample to pellet in a “new” rotor. The formula for this determination is as follows:
  • Where:
    T1 = Time to pellet in the “new” rotor
    T2 = Time to pellet in the “old” rotor
    K1 = K-factor of the “new” rotor
    K2 = K-factor of the “old” rotor
    Example of a rotor conversion:
    Old Rotor (Beckman® JA-10) New Rotor (Sorvall® SLC-1500)
    T2 = 20 min; K2 = 3610 T1 = ( ? ) min; K1 = 1676
    Old Pelleting Time = 20 min New Pelleting Time = 9.2 min
VII. References and suggested readings.
  1. Biological Centrifugation, by D. Rickwood, J.M. Graham (2001). Springer Verlag; ISBN: 0387915761
  2. Subcellular Fractionation: A Practical Approach, by John M. Graham (Editor) and D. Rickwood (Editor) (1997). Oxford Univ Press. ISBN: 0199634947
  3. Centrifugation: Essential Data, by David Rickwood, T. Ford, Jens Steensgaard (1994). 128 pages. John Wiley & Son Ltd. ISBN: 0471942715
  4. Centrifugation : A Practical Approach, by David Rickwood, (Editor) (1992) ASIN: 090414755X.
  5. An Introduction to Centrifugation, by TC. Ford and J.M. Graham (1991). 118 pages. BIOS Scientific Publishers, Ltd. ISBN 1 872748 40 6
Disclaimer: Masterflex products are not approved or intended for, and should not be used for medical, clinical, surgical or other patient-oriented applications.