PHARMED Tubing
Advantages:
  • Great for tissue and cell work—nontoxic and nonhemolytic.
  • Long service life minimizes risk of fluid exposure; reduces tubing costs and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids.
  • Heat sealable, bondable, and formable.
  • Extremely low gas permeability.
Limitations:
  • Potential leaching of USP mineral oil or blend material.
Application Suitability:
Acids: Good
Alkalies: Good
Organic solvents: Not recommended
Pressure: Good
Vacuum: Excellent
Viscous fluids: Good
Sterile fluids: Excellent
Physical Characteristics and Compostion:
  • Thermoplastic elastomer.
  • Polypropylene-based material with USP mineral oil.
  • Excellent tensile strength.
  • Firm (stiff) material.
  • Opaque, beige.
Temperature Range:
-75 to 275°F (-59 to 135°C)
Meets Classifications:
  • USP Class VI
  • FDA 21 CFR 177.2600
  • NSF listed (Standard 51)
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
  CO2: 1200
  H2: -
  O2: 200
  N2: 80
Cleaning/Sterilization:
  • Sterilize by ethylene oxide (ETO), autoclave or gamma irradiation up to 2.5 Mrad.
  • Repeated autoclaving will not affect overall life.